Assay Details
Gene Symbol: RAB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005571763.2 | XP_005571820.1 | 4 - 5 | 638 | 69 | |
XM_015431354.1 | XP_015286840.1 | 4 - 5 | 513 | 69 | ||
XM_015431355.1 | XP_015286841.1 | 4 - 5 | 419 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
