Assay Details
| Gene Symbol: MYZAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559641.2 | XP_005559698.1 | 3 - 4 | 447 | 82 | |
| XM_005559643.2 | XP_005559700.1 | 3 - 4 | 447 | 82 | ||
More Information
Set Membership: |
|

MYZAP
Genomic Map