Assay Details
Gene Symbol: CASC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005584075.2 | XP_005584132.1 | 8 - 9 | 1649 | 78 | |
XM_005584076.2 | XP_005584133.1 | 8 - 9 | 1653 | 78 | ||
XM_015438455.1 | XP_015293941.1 | 8 - 9 | 1653 | 78 | ||
GenBank mRNA | AB168159.1 | - | 4 - 5 | 1006 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
