Assay Details
| Gene Symbol: UGT3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556719.2 | XP_005556776.1 | 6 - 7 | 1681 | 87 | |
| XM_015451273.1 | XP_015306759.1 | 5 - 6 | 1156 | 87 | ||
| GenBank mRNA | AB169104.1 | - | 5 - 6 | 1150 | 87 | |
More Information
Set Membership: |
|

UGT3A1
Genomic Map