Assay Details
| Gene Symbol: POP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572407.2 | XP_005572464.1 | 2 - 3 | 236 | 71 | |
| XM_005572408.2 | XP_005572465.1 | 2 - 3 | 237 | 71 | ||
| XM_005572409.2 | XP_005572466.1 | 2 - 3 | 237 | 71 | ||
More Information
Set Membership: |
|

POP5
Genomic Map