Assay Details
Gene Symbol: TFB1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283474.1 | NP_001270403.1 | 1 - 2 | 186 | 73 | |
XM_015448831.1 | XP_015304317.1 | 1 - 2 | 213 | 73 | ||
XM_015448832.1 | XP_015304318.1 | 1 - 2 | 213 | 73 | ||
XM_015448833.1 | XP_015304319.1 | 1 - 2 | 213 | 73 | ||
GenBank mRNA | AB220391.1 | - | 1 - 2 | 186 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
