Assay Details
Gene Symbol: DIABLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005572481.2 | XP_005572538.1 | 4 - 5 | 498 | 79 | |
XM_005572482.2 | XP_005572539.1 | 4 - 5 | 432 | 79 | ||
XM_005572484.2 | XP_005572541.1 | 4 - 5 | 458 | 79 | ||
XM_005572485.2 | XP_005572542.1 | 3 - 4 | 319 | 79 | ||
GenBank mRNA | AB170199.1 | - | 4 - 5 | 430 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
