Assay Details
| Gene Symbol: RMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587027.2 | XP_005587084.1 | 14 - 15 | 1444 | 66 | |
| XM_015440103.1 | XP_015295589.1 | 14 - 15 | 1379 | 66 | ||
| XM_015440104.1 | XP_015295590.1 | 14 - 15 | 1329 | 66 | ||
| XM_015440105.1 | XP_015295591.1 | 14 - 15 | 1444 | 66 | ||
More Information
Set Membership: |
|

RMC1
Genomic Map