Assay Details
| Gene Symbol: HAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588836.2 | XP_005588893.1 | 1 - 2 | 325 | 65 | |
| GenBank mRNA | EU076443.1 | - | 1 - 2 | 131 | 65 | |
More Information
Set Membership: |
|

HAMP
Genomic Map