Assay Details
Gene Symbol: DELE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005558078.2 | XP_005558135.1 | 2 - 3 | 313 | 85 | |
XM_005558079.2 | XP_005558136.1 | 2 - 3 | 313 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
