Assay Details
| Gene Symbol: PTBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587328.2 | XP_005587385.1 | 11 - 12 | 1739 | 90 | |
| XM_005587329.2 | XP_005587386.1 | 11 - 12 | 1263 | 90 | ||
| XM_005587331.2 | XP_005587388.1 | 10 - 11 | 1661 | 90 | ||
| XM_005587332.2 | XP_005587389.1 | 10 - 11 | 1185 | 90 | ||
| XM_005587333.2 | XP_005587390.1 | 10 - 11 | 1215 | 90 | ||
More Information
Set Membership: |
|

PTBP1
Genomic Map