Assay Details
Gene Symbol: STK26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319600.1 | NP_001306529.1 | 6 - 7 | 781 | 118 | |
XM_015444139.1 | XP_015299625.1 | 7 - 8 | 931 | 118 | ||
XM_015444141.1 | XP_015299627.1 | 7 - 8 | 934 | 118 | ||
XM_015444142.1 | XP_015299628.1 | 7 - 8 | 934 | 118 | ||
XM_015444143.1 | XP_015299629.1 | 6 - 7 | 894 | 118 | ||
XM_015444144.1 | XP_015299630.1 | 5 - 6 | 640 | 118 | ||
GenBank mRNA | AB168500.1 | - | 6 - 7 | 781 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
