Assay Details
Gene Symbol: MGMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566748.2 | XP_005566805.1 | 2 - 3 | 243 | 93 | |
XM_005566749.2 | XP_005566806.1 | 2 - 3 | 243 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
