Assay Details
Gene Symbol: NSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285058.1 | NP_001271987.1 | 2 - 3 | 316 | 71 | |
XM_005554461.2 | XP_005554518.1 | 2 - 3 | 601 | 71 | ||
XM_005554462.2 | XP_005554519.1 | 2 - 3 | 716 | 71 | ||
GenBank mRNA | AB169490.1 | - | 2 - 3 | 236 | 71 | |
AB170586.1 | - | 2 - 3 | 236 | 71 | ||
AB174648.1 | - | 2 - 3 | 316 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
