Assay Details
| Gene Symbol: CDA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544549.2 | XP_005544606.1 | 1 - 2 | 332 | 98 | |
| XM_005544550.2 | XP_005544607.1 | 1 - 2 | 315 | 98 | ||
More Information
Set Membership: |
|

CDA
Genomic Map