Assay Details
Gene Symbol: CD59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283527.1 | NP_001270456.1 | 3 - 4 | 229 | 87 | |
XM_015434110.1 | XP_015289596.1 | 4 - 5 | 329 | 87 | ||
GenBank mRNA | AB072017.1 | - | 2 - 3 | 169 | 87 | |
AB072018.1 | - | 2 - 3 | 169 | 87 | ||
AB169517.1 | - | 3 - 4 | 253 | 87 | ||
AB170328.1 | - | 3 - 4 | 239 | 87 | ||
AB171356.1 | - | 3 - 4 | 229 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
