Assay Details
| Gene Symbol: PLAC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554911.2 | XP_005554968.1 | 2 - 3 | 292 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLAC8| Gene Symbol: PLAC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554911.2 | XP_005554968.1 | 2 - 3 | 292 | 77 | |
Set Membership: |
|