Assay Details
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556604.2 | XP_005556661.1 | 5 - 6 | 741 | 90 | |
| XM_005556607.2 | XP_005556664.1 | 5 - 6 | 495 | 90 | ||
| XM_015451220.1 | XP_015306706.1 | 4 - 5 | 599 | 90 | ||
More Information
Set Membership: |
|

OTULIN
Genomic Map