Assay Details
| Gene Symbol: SRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557984.2 | XP_005558041.1 | 4 - 5 | 1783 | 102 | |
| XM_005557985.2 | XP_005558042.1 | 2 - 3 | 1773 | 102 | ||
More Information
Set Membership: |
|

SRA1
Genomic Map