Assay Details
Gene Symbol: LOC102142263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005540677.1 | XP_005540734.1 | 2 - 3 | 372 | 62 | |
XM_005540678.2 | XP_005540735.1 | 2 - 3 | 370 | 62 | ||
XM_005540679.1 | XP_005540736.1 | 2 - 3 | 372 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
