Assay Details
Gene Symbol: ADTRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005554034.2 | XP_005554091.1 | 2 - 3 | 1519 | 72 | |
XM_015449897.1 | XP_015305383.1 | 3 - 4 | 668 | 72 | ||
XM_015449898.1 | XP_015305384.1 | 2 - 3 | 426 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
