Assay Details
| Gene Symbol: LHPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566682.2 | XP_005566739.1 | 4 - 5 | 607 | 66 | |
| XM_005566683.2 | XP_005566740.1 | 4 - 5 | 607 | 66 | ||
| XM_005566684.2 | XP_005566741.1 | 4 - 5 | 607 | 66 | ||
| XM_005566685.2 | XP_005566742.1 | 4 - 5 | 607 | 66 | ||
| XM_005566686.2 | XP_005566743.1 | 4 - 5 | 607 | 66 | ||
| XM_015456830.1 | XP_015312316.1 | 4 - 5 | 607 | 66 | ||
More Information
Set Membership: |
|

LHPP
Genomic Map