Assay Details
| Gene Symbol: AP3S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557554.2 | XP_005557611.1 | 1 - 2 | 509 | 61 | |
| XM_005557556.2 | XP_005557613.1 | 1 - 2 | 509 | 61 | ||
| XM_015451808.1 | XP_015307294.1 | 1 - 2 | 509 | 61 | ||
| XM_015451809.1 | XP_015307295.1 | 1 - 2 | 344 | 61 | ||
More Information
Set Membership: |
|

AP3S1
Genomic Map