Assay Details
Gene Symbol: LOC102137053 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287711.1 | NP_001274640.1 | 5 - 6 | 485 | 99 | |
XM_005567098.2 | XP_005567155.1 | 5 - 6 | 702 | 99 | ||
GenBank mRNA | AB125163.1 | - | 5 - 6 | 543 | 99 | |
AB169480.1 | - | 5 - 6 | 485 | 99 | ||
AB179491.1 | - | 5 - 6 | 485 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
