Assay Details
Gene Symbol: NDUFAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284603.1 | NP_001271532.1 | 1 - 2 | 223 | 81 | |
GenBank mRNA | AB169788.1 | - | 1 - 2 | 223 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
