Assay Details
Gene Symbol: MAP3K7CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005548865.2 | XP_005548922.1 | 3 - 4 | 610 | 79 | |
XM_015446936.1 | XP_015302422.1 | 2 - 3 | 211 | 79 | ||
XM_015446938.1 | XP_015302424.1 | 2 - 3 | 317 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015446936.1, XM_015446938.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
