Assay Details
| Gene Symbol: RBM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560822.2 | XP_005560879.1 | 9 - 10 | 1060 | 102 | |
| XM_005560823.2 | XP_005560880.1 | 9 - 10 | 1059 | 102 | ||
| XM_005560824.2 | XP_005560881.1 | 9 - 10 | 952 | 102 | ||
| XM_005560826.2 | XP_005560883.1 | 8 - 9 | 1012 | 102 | ||
| XM_015453572.1 | XP_015309058.1 | 8 - 9 | 1011 | 102 | ||
More Information
Set Membership: |
|

RBM23
Genomic Map