Assay Details
| Gene Symbol: RNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586955.2 | XP_005587012.1 | 7 - 8 | 1094 | 119 | |
| XM_005586956.2 | XP_005587013.1 | 6 - 7 | 907 | 119 | ||
| XM_005586957.2 | XP_005587014.1 | 6 - 7 | 749 | 119 | ||
More Information
Set Membership: |
|

RNF138
Genomic Map