Assay Details
Gene Symbol: RAB11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005559836.2 | XP_005559893.1 | 4 - 5 | 801 | 84 | |
GenBank mRNA | AB169729.1 | - | 4 - 5 | 610 | 84 | |
AB173395.1 | - | 2 - 3 | 271 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
