Assay Details
| Gene Symbol: EDARADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283135.1 | NP_001270064.1 | 6 - 7 | 464 | 104 | |
| XM_015446193.1 | XP_015301679.1 | 4 - 5 | 379 | 104 | ||
| XM_015446226.1 | XP_015301712.1 | 4 - 5 | 716 | 104 | ||
| XM_015446258.1 | XP_015301744.1 | 6 - 7 | 891 | 104 | ||
| XM_015446291.1 | XP_015301777.1 | 5 - 6 | 810 | 104 | ||
| GenBank mRNA | AB072753.1 | - | 6 - 7 | 464 | 104 | |
More Information
Set Membership: |
|

EDARADD
Genomic Map