Assay Details
| Gene Symbol: CEP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545291.2 | XP_005545348.1 | 2 - 3 | 562 | 113 | |
| XM_005545293.2 | XP_005545350.1 | 2 - 3 | 295 | 113 | ||
More Information
Set Membership: |
|

CEP19
Genomic Map