Assay Details
| Gene Symbol: PVRIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549224.2 | XP_005549281.1 | 4 - 5 | 1985 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PVRIG| Gene Symbol: PVRIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549224.2 | XP_005549281.1 | 4 - 5 | 1985 | 77 | |
Set Membership: |
|