Assay Details
| Gene Symbol: GPAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563168.2 | XP_005563225.1 | 4 - 5 | 1508 | 67 | |
| XM_005563169.1 | XP_005563226.1 | 4 - 5 | 1844 | 67 | ||
| XM_005563170.2 | XP_005563227.1 | 3 - 4 | 444 | 67 | ||
| XM_015454711.1 | XP_015310197.1 | 4 - 5 | 1399 | 67 | ||
| XM_015454712.1 | XP_015310198.1 | 4 - 5 | 1397 | 67 | ||
| XM_015454713.1 | XP_015310199.1 | 4 - 5 | 1397 | 67 | ||
| XM_015454714.1 | XP_015310200.1 | 4 - 5 | 1397 | 67 | ||
| XM_015454715.1 | XP_015310201.1 | 4 - 5 | 1968 | 67 | ||
More Information
Set Membership: |
|

GPAT4
Genomic Map