Assay Details
| Gene Symbol: CARHSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591218.2 | XP_005591275.1 | 2 - 3 | 506 | 71 | |
| XM_005591219.2 | XP_005591276.1 | 2 - 3 | 276 | 71 | ||
| XM_005591220.2 | XP_005591277.1 | 2 - 3 | 539 | 71 | ||
| XM_015442459.1 | XP_015297945.1 | 2 - 3 | 406 | 71 | ||
More Information
Set Membership: |
|

CARHSP1
Genomic Map