Assay Details
| Gene Symbol: ARF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570734.2 | XP_005570791.1 | 3 - 4 | 705 | 107 | |
| XM_005570735.2 | XP_005570792.1 | 2 - 3 | 476 | 107 | ||
| XM_015430868.1 | XP_015286354.1 | 2 - 3 | 260 | 107 | ||
| GenBank mRNA | AB220383.1 | - | 2 - 3 | 394 | 107 | |
More Information
Set Membership: |
|

ARF3
Genomic Map