Assay Details
Gene Symbol: ARL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566317.2 | XP_005566374.1 | 3 - 4 | 424 | 114 | |
XM_015456630.1 | XP_015312116.1 | 3 - 4 | 424 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
