Assay Details
Gene Symbol: DDX39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588244.2 | XP_005588301.1 | 7 - 8 | 999 | 55 | |
XM_005588245.2 | XP_005588302.1 | 7 - 8 | 2007 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
