Assay Details
| Gene Symbol: MFAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559389.1 | XP_005559446.1 | 7 - 8 | 1228 | 123 | |
| GenBank mRNA | AB168920.1 | - | 7 - 8 | 1165 | 123 | |
More Information
Set Membership: |
|

MFAP1
Genomic Map