Assay Details
| Gene Symbol: EFEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319411.1 | NP_001306340.1 | 4 - 5 | 592 | 83 | |
| XM_015433644.1 | XP_015289130.1 | 4 - 5 | 681 | 83 | ||
| GenBank mRNA | AY312415.1 | - | 4 - 5 | 592 | 83 | |
More Information
Set Membership: |
|

EFEMP1
Genomic Map