Assay Details
Gene Symbol: C4H6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553148.2 | XP_005553205.1 | 3 - 4 | 602 | 91 | |
XM_005553149.2 | XP_005553206.1 | 4 - 5 | 708 | 91 | ||
XM_005553150.2 | XP_005553207.1 | 5 - 6 | 730 | 91 | ||
XM_005553153.2 | XP_005553210.1 | 3 - 4 | 424 | 91 | ||
XM_015449474.1 | XP_015304960.1 | 5 - 6 | 775 | 91 | ||
XM_015449478.1 | XP_015304964.1 | 3 - 4 | 365 | 91 | ||
GenBank mRNA | AB171021.1 | - | 4 - 5 | 597 | 91 | |
AB171198.1 | - | 3 - 4 | 480 | 91 | ||
AB171475.1 | - | 5 - 6 | 730 | 91 | ||
AB171487.1 | - | 155 | 91 |
