Assay Details
Gene Symbol: CAAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581590.2 | XP_005581647.1 | 4 - 5 | 1125 | 103 | |
XM_005581591.2 | XP_005581648.1 | 4 - 5 | 934 | 103 | ||
GenBank mRNA | AB168912.1 | - | 5 - 6 | 561 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
