Assay Details
| Gene Symbol: PDIA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559378.2 | XP_005559435.1 | 1 - 2 | 391 | 69 | |
| XM_005559379.2 | XP_005559436.1 | 1 - 2 | 391 | 69 | ||
More Information
Set Membership: |
|

PDIA3
Genomic Map