Assay Details
| Gene Symbol: ADH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555523.2 | XP_005555580.1 | 3 - 4 | 405 | 93 | |
| XM_005555524.2 | XP_005555581.1 | 3 - 4 | 838 | 93 | ||
More Information
Set Membership: |
|

ADH7
Genomic Map