Assay Details
| Gene Symbol: AGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283634.1 | NP_001270563.1 | 2 - 3 | 895 | 64 | |
| XM_015443695.1 | XP_015299181.1 | 1 - 2 | 1383 | 64 | ||
| GenBank mRNA | AB170313.1 | - | 2 - 3 | 895 | 64 | |
More Information
Set Membership: |
|

AGT
Genomic Map