Assay Details
| Gene Symbol: SNAPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284877.1 | NP_001271806.1 | 3 - 4 | 478 | 95 | |
| XM_015436482.1 | XP_015291968.1 | 3 - 4 | 1048 | 95 | ||
| GenBank mRNA | AB169044.1 | - | 3 - 4 | 478 | 95 | |
More Information
Set Membership: |
|

SNAPC3
Genomic Map