Assay Details
| Gene Symbol: HM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568623.2 | XP_005568680.1 | 10 - 11 | 1103 | 87 | |
| XM_005568624.2 | XP_005568681.1 | 10 - 11 | 1103 | 87 | ||
| XM_005568625.2 | XP_005568682.1 | 7 - 8 | 1466 | 87 | ||
| XM_015457883.1 | XP_015313369.1 | 9 - 10 | 854 | 87 | ||
More Information
Set Membership: |
|

HM13
Genomic Map