Assay Details
Gene Symbol: BDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555579.2 | XP_005555636.1 | 8 - 9 | 732 | 105 | |
XM_015450713.1 | XP_015306199.1 | 9 - 10 | 777 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
