Assay Details
Gene Symbol: DDAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553487.2 | XP_005553544.1 | 4 - 5 | 751 | 57 | |
XM_005553488.2 | XP_005553545.1 | 4 - 5 | 848 | 57 | ||
XM_015449612.1 | XP_015305098.1 | 3 - 4 | 1589 | 57 | ||
XM_015449613.1 | XP_015305099.1 | 4 - 5 | 760 | 57 | ||
XM_015449614.1 | XP_015305100.1 | 4 - 5 | 776 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
