Assay Details
| Gene Symbol: POM121C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549354.2 | XP_005549411.1 | 11 - 12 | 3503 | 114 | |
| XM_015447209.1 | XP_015302695.1 | 11 - 12 | 3065 | 114 | ||
More Information
Set Membership: |
|

POM121C
Genomic Map