Assay Details
| Gene Symbol: SYNGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567237.2 | XP_005567294.1 | 2 - 3 | 1773 | 75 | |
| XM_005567238.2 | XP_005567295.1 | 2 - 3 | 1787 | 75 | ||
| XM_005567239.2 | XP_005567296.1 | 2 - 3 | 1761 | 75 | ||
| XM_005567242.2 | XP_005567299.1 | 2 - 3 | 2976 | 75 | ||
More Information
Set Membership: |
|

SYNGR1
Genomic Map